Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600909 MIM: 612412 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LSS - lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001438.2 | Intron | NP_001001438.1 | ||||
NM_001145436.1 | Intron | NP_001138908.1 | ||||
NM_001145437.1 | Intron | NP_001138909.1 | ||||
NM_002340.5 | Intron | NP_002331.3 | ||||
XM_006724004.3 | Intron | XP_006724067.1 | ||||
XM_011529564.2 | Intron | XP_011527866.1 | ||||
XM_011529565.1 | Intron | XP_011527867.1 | ||||
XM_011529566.2 | Intron | XP_011527868.1 | ||||
XM_011529567.2 | Intron | XP_011527869.1 | ||||
XM_017028346.1 | Intron | XP_016883835.1 | ||||
XM_017028347.1 | Intron | XP_016883836.1 | ||||
XM_017028348.1 | Intron | XP_016883837.1 |
SPATC1L - spermatogenesis and centriole associated 1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |