Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1211L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIAA1211L - KIAA1211 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207362.2 | Intron | NP_997245.2 | ||||
XM_005263923.3 | Intron | XP_005263980.1 | ||||
XM_005263924.3 | Intron | XP_005263981.1 | ||||
XM_005263926.3 | Intron | XP_005263983.1 | ||||
XM_011511093.1 | Intron | XP_011509395.1 | ||||
XM_011511094.1 | Intron | XP_011509396.1 | ||||
XM_011511095.2 | Intron | XP_011509397.1 | ||||
XM_011511096.1 | Intron | XP_011509398.1 | ||||
XM_017003971.1 | Intron | XP_016859460.1 | ||||
XM_017003972.1 | Intron | XP_016859461.1 | ||||
XM_017003973.1 | Intron | XP_016859462.1 |