Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KIAA1211 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA1211 - KIAA1211 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020722.1 | Intron | NP_065773.1 | ||||
XM_005265752.2 | Intron | XP_005265809.1 | ||||
XM_005265755.2 | Intron | XP_005265812.1 | ||||
XM_005265756.2 | Intron | XP_005265813.1 | ||||
XM_006714045.2 | Intron | XP_006714108.1 | ||||
XM_011534397.2 | Intron | XP_011532699.1 | ||||
XM_017008473.1 | Intron | XP_016863962.1 | ||||
XM_017008474.1 | Intron | XP_016863963.1 |