Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607582 MIM: 607581 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC22A6 - solute carrier family 22 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A8 - solute carrier family 22 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184732.1 | Intron | NP_001171661.1 | ||||
NM_001184733.1 | Intron | NP_001171662.1 | ||||
NM_001184736.1 | Intron | NP_001171665.1 | ||||
NM_004254.3 | Intron | NP_004245.2 | ||||
XM_011545364.1 | Intron | XP_011543666.1 |
Set Membership: |
HapMap |