Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138248 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GRIA1 - glutamate ionotropic receptor AMPA type subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000827.3 | Intron | NP_000818.2 | ||||
NM_001114183.1 | Intron | NP_001107655.1 | ||||
NM_001258019.1 | Intron | NP_001244948.1 | ||||
NM_001258020.1 | Intron | NP_001244949.1 | ||||
NM_001258021.1 | Intron | NP_001244950.1 | ||||
NM_001258022.1 | Intron | NP_001244951.1 | ||||
NM_001258023.1 | Intron | NP_001244952.1 | ||||
XM_011537635.2 | Intron | XP_011535937.1 | ||||
XM_017009392.1 | Intron | XP_016864881.1 | ||||
XM_017009393.1 | Intron | XP_016864882.1 |
Set Membership: |
HapMap |