Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606021 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRAME PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PRAME - preferentially expressed antigen in melanoma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291715.1 | 1788 | Silent Mutation | TCC,TCT | S,S 421 | NP_001278644.1 | |
NM_001291716.1 | 1788 | Silent Mutation | TCC,TCT | S,S 421 | NP_001278645.1 | |
NM_001291717.1 | 1788 | Silent Mutation | TCC,TCT | S,S 405 | NP_001278646.1 | |
NM_001291719.1 | 1788 | Silent Mutation | TCC,TCT | S,S 405 | NP_001278648.1 | |
NM_001318126.1 | 1788 | Silent Mutation | TCC,TCT | S,S 405 | NP_001305055.1 | |
NM_001318127.1 | 1788 | Silent Mutation | TCC,TCT | S,S 405 | NP_001305056.1 | |
NM_006115.4 | 1788 | Silent Mutation | TCC,TCT | S,S 421 | NP_006106.1 | |
NM_206953.2 | 1788 | Silent Mutation | TCC,TCT | S,S 421 | NP_996836.1 | |
NM_206954.2 | 1788 | Silent Mutation | TCC,TCT | S,S 421 | NP_996837.1 | |
NM_206955.2 | 1788 | Silent Mutation | TCC,TCT | S,S 421 | NP_996838.1 | |
NM_206956.2 | 1788 | Silent Mutation | TCC,TCT | S,S 421 | NP_996839.1 | |
XM_011530034.2 | 1788 | Silent Mutation | TCC,TCT | S,S 405 | XP_011528336.1 |