Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610408 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC15A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC15A3 - solute carrier family 15 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016582.2 | 1713 | Silent Mutation | GGC,GGT | G,G 493 | NP_057666.1 | |
XM_011545095.2 | 1713 | Silent Mutation | GGC,GGT | G,G 493 | XP_011543397.1 |
TMEM132A - transmembrane protein 132A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017870.3 | 1713 | Intron | NP_060340.2 | |||
NM_178031.2 | 1713 | Intron | NP_821174.1 | |||
XM_017017951.1 | 1713 | Intron | XP_016873440.1 | |||
XM_017017952.1 | 1713 | Intron | XP_016873441.1 |