Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ERC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERC2 - ELKS/RAB6-interacting/CAST family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015576.2 | Intron | NP_056391.1 | ||||
XM_017006138.1 | Intron | XP_016861627.1 | ||||
XM_017006139.1 | Intron | XP_016861628.1 | ||||
XM_017006140.1 | Intron | XP_016861629.1 | ||||
XM_017006141.1 | Intron | XP_016861630.1 | ||||
XM_017006142.1 | Intron | XP_016861631.1 | ||||
XM_017006143.1 | Intron | XP_016861632.1 | ||||
XM_017006144.1 | Intron | XP_016861633.1 | ||||
XM_017006145.1 | Intron | XP_016861634.1 | ||||
XM_017006146.1 | Intron | XP_016861635.1 | ||||
XM_017006147.1 | Intron | XP_016861636.1 | ||||
XM_017006148.1 | Intron | XP_016861637.1 | ||||
XM_017006149.1 | Intron | XP_016861638.1 | ||||
XM_017006150.1 | Intron | XP_016861639.1 | ||||
XM_017006151.1 | Intron | XP_016861640.1 | ||||
XM_017006152.1 | Intron | XP_016861641.1 | ||||
XM_017006153.1 | Intron | XP_016861642.1 | ||||
XM_017006154.1 | Intron | XP_016861643.1 | ||||
XM_017006155.1 | Intron | XP_016861644.1 | ||||
XM_017006156.1 | Intron | XP_016861645.1 | ||||
XM_017006157.1 | Intron | XP_016861646.1 | ||||
XM_017006158.1 | Intron | XP_016861647.1 | ||||
XM_017006159.1 | Intron | XP_016861648.1 |
ERC2-IT1 - ERC2 intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |