Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616706 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PARP11 - poly(ADP-ribose) polymerase family member 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286521.1 | Intron | NP_001273450.1 | ||||
NM_001286522.1 | Intron | NP_001273451.1 | ||||
NM_020367.5 | Intron | NP_065100.2 | ||||
XM_005253711.4 | Intron | XP_005253768.1 | ||||
XM_011520970.1 | Intron | XP_011519272.1 | ||||
XM_011520971.2 | Intron | XP_011519273.1 | ||||
XM_011520973.2 | Intron | XP_011519275.1 | ||||
XM_017019670.1 | Intron | XP_016875159.1 | ||||
XM_017019671.1 | Intron | XP_016875160.1 |