Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610465 MIM: 602178 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ACSF2 - acyl-CoA synthetase family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288968.1 | 1920 | Intron | NP_001275897.1 | |||
NM_001288969.1 | 1920 | Intron | NP_001275898.1 | |||
NM_001288970.1 | 1920 | Intron | NP_001275899.1 | |||
NM_001288971.1 | 1920 | Intron | NP_001275900.1 | |||
NM_001288972.1 | 1920 | Intron | NP_001275901.1 | |||
NM_025149.5 | 1920 | Intron | NP_079425.3 | |||
XM_006722110.3 | 1920 | Intron | XP_006722173.1 | |||
XM_011525294.1 | 1920 | Intron | XP_011523596.1 |
CHAD - chondroadherin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267.2 | 1920 | Silent Mutation | CCC,CCT | P,P 284 | NP_001258.2 | |
XM_011524214.2 | 1920 | Silent Mutation | CCC,CCT | P,P 365 | XP_011522516.1 |
Set Membership: |
HapMap |