Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609906 MIM: 611461 | ||||||||||||||||||||
Literature Links: |
EFS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EFS - embryonal Fyn-associated substrate | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277174.1 | 1637 | Silent Mutation | GGC,GGT | G,G 343 | NP_001264103.1 | |
NM_005864.3 | 1637 | Silent Mutation | GGC,GGT | G,G 512 | NP_005855.1 | |
NM_032459.2 | 1637 | Silent Mutation | GGC,GGT | G,G 419 | NP_115835.1 | |
XM_005267256.1 | 1637 | Silent Mutation | GGC,GGT | G,G 482 | XP_005267313.1 |
SLC22A17 - solute carrier family 22 member 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |