Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606942 MIM: 604800 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPE PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
COPE - coatomer protein complex subunit epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007263.3 | Intron | NP_009194.2 | ||||
NM_199442.1 | Intron | NP_955474.1 | ||||
NM_199444.1 | Intron | NP_955476.1 | ||||
XM_011527655.1 | Intron | XP_011525957.1 | ||||
XM_011527656.1 | Intron | XP_011525958.1 |
DDX49 - DEAD-box helicase 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019070.4 | Intron | NP_061943.2 | ||||
XM_011528083.1 | Intron | XP_011526385.1 | ||||
XM_011528084.2 | Intron | XP_011526386.1 |
HOMER3 - homer scaffolding protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |