Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601826 MIM: 610570 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKD PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DGKD - diacylglycerol kinase delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP40 - ubiquitin specific peptidase 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018218.2 | 4165 | UTR 3 | NP_060688.1 | |||
XM_006712612.3 | 4165 | UTR 3 | XP_006712675.1 | |||
XM_011511396.2 | 4165 | UTR 3 | XP_011509698.1 | |||
XM_011511397.2 | 4165 | UTR 3 | XP_011509699.1 | |||
XM_011511398.2 | 4165 | Intron | XP_011509700.1 | |||
XM_011511399.2 | 4165 | UTR 3 | XP_011509701.1 | |||
XM_011511400.2 | 4165 | UTR 3 | XP_011509702.1 | |||
XM_011511401.2 | 4165 | Intron | XP_011509703.1 | |||
XM_017004426.1 | 4165 | UTR 3 | XP_016859915.1 | |||
XM_017004427.1 | 4165 | UTR 3 | XP_016859916.1 | |||
XM_017004428.1 | 4165 | UTR 3 | XP_016859917.1 |
Set Membership: |
HapMap |