Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C3orf18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C3orf18 - chromosome 3 open reading frame 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171740.2 | Intron | NP_001165211.1 | ||||
NM_001171741.2 | Intron | NP_001165212.1 | ||||
NM_001171743.2 | Intron | NP_001165214.2 | ||||
NM_016210.4 | Intron | NP_057294.2 | ||||
XM_011533781.2 | Intron | XP_011532083.1 | ||||
XM_011533782.2 | Intron | XP_011532084.1 | ||||
XM_011533783.2 | Intron | XP_011532085.1 | ||||
XM_011533784.2 | Intron | XP_011532086.1 | ||||
XM_011533785.2 | Intron | XP_011532087.1 | ||||
XM_011533788.1 | Intron | XP_011532090.1 | ||||
XM_011533789.1 | Intron | XP_011532091.1 | ||||
XM_011533790.1 | Intron | XP_011532092.1 | ||||
XM_017006545.1 | Intron | XP_016862034.1 | ||||
XM_017006546.1 | Intron | XP_016862035.1 | ||||
XM_017006547.1 | Intron | XP_016862036.1 |
HEMK1 - HemK methyltransferase family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317851.1 | Intron | NP_001304780.1 | ||||
NM_016173.4 | Intron | NP_057257.1 | ||||
XM_005265218.3 | Intron | XP_005265275.1 | ||||
XM_011533806.2 | Intron | XP_011532108.1 | ||||
XM_011533807.1 | Intron | XP_011532109.1 | ||||
XM_011533808.1 | Intron | XP_011532110.1 | ||||
XM_011533809.2 | Intron | XP_011532111.1 | ||||
XM_011533810.2 | Intron | XP_011532112.1 | ||||
XM_011533812.2 | Intron | XP_011532114.1 | ||||
XM_011533813.2 | Intron | XP_011532115.1 | ||||
XM_011533814.2 | Intron | XP_011532116.1 |