Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162341 MIM: 610902 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMBR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NMBR - neuromedin B receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324307.1 | 238 | Intron | NP_001311236.1 | |||
NM_001324308.1 | 238 | Intron | NP_001311237.1 | |||
NM_002511.3 | 238 | Intron | NP_002502.2 | |||
XM_011535846.1 | 238 | Intron | XP_011534148.1 | |||
XM_017010901.1 | 238 | Intron | XP_016866390.1 | |||
XM_017010902.1 | 238 | Intron | XP_016866391.1 |
VTA1 - vesicle trafficking 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286371.1 | 238 | Missense Mutation | AGT,TGT | S,C 38 | NP_001273300.1 | |
NM_001286372.1 | 238 | Silent Mutation | ACA,ACT | T,T 11 | NP_001273301.1 | |
NM_016485.4 | 238 | Missense Mutation | AGT,TGT | S,C 38 | NP_057569.2 |