Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
50 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2U PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UBE2U - ubiquitin conjugating enzyme E2 U (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152489.1 | Intron | NP_689702.1 | ||||
XM_005270498.4 | Intron | XP_005270555.1 | ||||
XM_006710379.3 | Intron | XP_006710442.1 | ||||
XM_011540764.2 | Intron | XP_011539066.1 | ||||
XM_017000379.1 | Intron | XP_016855868.1 | ||||
XM_017000380.1 | Intron | XP_016855869.1 | ||||
XM_017000381.1 | Intron | XP_016855870.1 | ||||
XM_017000382.1 | Intron | XP_016855871.1 | ||||
XM_017000383.1 | Intron | XP_016855872.1 | ||||
XM_017000384.1 | Intron | XP_016855873.1 | ||||
XM_017000385.1 | Intron | XP_016855874.1 |