Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603153 MIM: 616344 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRIX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRIX1 - BRX1, biogenesis of ribosomes | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAD1 - RAD1 checkpoint DNA exonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002853.3 | 1970 | UTR 3 | NP_002844.1 |
TTC23L - tetratricopeptide repeat domain 23 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |