Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616597 MIM: 176889 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL8A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARL8A - ADP ribosylation factor like GTPase 8A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPN7 - protein tyrosine phosphatase, non-receptor type 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199797.1 | 2088 | UTR 3 | NP_001186726.1 | |||
NM_002832.3 | 2088 | UTR 3 | NP_002823.3 | |||
NM_080588.2 | 2088 | UTR 3 | NP_542155.1 | |||
XM_011509819.1 | 2088 | UTR 3 | XP_011508121.1 | |||
XM_011509820.2 | 2088 | UTR 3 | XP_011508122.1 | |||
XM_011509821.1 | 2088 | UTR 3 | XP_011508123.1 | |||
XM_011509822.2 | 2088 | UTR 3 | XP_011508124.1 | |||
XM_011509823.1 | 2088 | Intron | XP_011508125.1 | |||
XM_011509824.1 | 2088 | UTR 3 | XP_011508126.1 | |||
XM_017001935.1 | 2088 | UTR 3 | XP_016857424.1 | |||
XM_017001936.1 | 2088 | UTR 3 | XP_016857425.1 | |||
XM_017001937.1 | 2088 | UTR 3 | XP_016857426.1 |