Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605874 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNK9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNK9 - potassium two pore domain channel subfamily K member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282534.1 | Intron | NP_001269463.1 | ||||
XM_011517101.2 | Intron | XP_011515403.1 | ||||
XM_011517102.2 | Intron | XP_011515404.1 | ||||
XM_011517103.2 | Intron | XP_011515405.1 | ||||
XM_017013530.1 | Intron | XP_016869019.1 | ||||
XM_017013531.1 | Intron | XP_016869020.1 |
Set Membership: |
HapMap |