Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM177B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM177B - family with sequence similarity 177 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324080.1 | Intron | NP_001311009.1 | ||||
NM_207468.2 | Intron | NP_997351.2 | ||||
XM_017001279.1 | Intron | XP_016856768.1 | ||||
XM_017001280.1 | Intron | XP_016856769.1 | ||||
XM_017001281.1 | Intron | XP_016856770.1 | ||||
XM_017001282.1 | Intron | XP_016856771.1 | ||||
XM_017001283.1 | Intron | XP_016856772.1 | ||||
XM_017001284.1 | Intron | XP_016856773.1 | ||||
XM_017001285.1 | Intron | XP_016856774.1 |