Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601955 MIM: 611996 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDK7 - cyclin dependent kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324069.1 | Intron | NP_001310998.1 | ||||
NM_001324070.1 | Intron | NP_001310999.1 | ||||
NM_001324071.1 | Intron | NP_001311000.1 | ||||
NM_001324072.1 | Intron | NP_001311001.1 | ||||
NM_001324074.1 | Intron | NP_001311003.1 | ||||
NM_001324075.1 | Intron | NP_001311004.1 | ||||
NM_001324077.1 | Intron | NP_001311006.1 | ||||
NM_001324078.1 | Intron | NP_001311007.1 | ||||
NM_001799.3 | Intron | NP_001790.1 | ||||
XM_011543094.2 | Intron | XP_011541396.1 |
MRPS36 - mitochondrial ribosomal protein S36 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |