Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGTGTGGACAGGTGGGGGGCGCA[A/G]AGGACAGGGCTGGTCTGGAGCCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612920 | ||||||||||||||||||||
Literature Links: |
KRTAP10-11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRTAP10-11 - keratin associated protein 10-11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTAP12-3 - keratin associated protein 12-3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTAP12-4 - keratin associated protein 12-4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198698.1 | 17 | UTR 5 | NP_941971.1 |
TSPEAR - thrombospondin type laminin G domain and EAR repeats | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272037.1 | 17 | Intron | NP_001258966.1 | |||
NM_144991.2 | 17 | Intron | NP_659428.2 |