Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603191 MIM: 171860 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C21orf2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C21orf2 - chromosome 21 open reading frame 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271440.1 | 1207 | Intron | NP_001258369.1 | |||
NM_001271441.1 | 1207 | Silent Mutation | CCG,CCT | P,P 333 | NP_001258370.1 | |
NM_001271442.1 | 1207 | Intron | NP_001258371.1 | |||
NM_004928.2 | 1207 | Intron | NP_004919.1 | |||
XM_006724051.3 | 1207 | Intron | XP_006724114.1 | |||
XM_006724053.3 | 1207 | Intron | XP_006724116.1 | |||
XM_017028470.1 | 1207 | Intron | XP_016883959.1 | |||
XM_017028471.1 | 1207 | Intron | XP_016883960.1 | |||
XM_017028472.1 | 1207 | Intron | XP_016883961.1 |
PFKL - phosphofructokinase, liver type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |