Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 130620 | |||||||||||||||||||||||||||||
Literature Links: |
RPS14 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
RPS14 - ribosomal protein S14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025070.1 | 511 | Intron | NP_001020241.1 | |||
NM_001025071.1 | 511 | Intron | NP_001020242.1 | |||
NM_005617.3 | 511 | Intron | NP_005608.1 | |||
XM_017009684.1 | 511 | Missense Mutation | CCT,CGT | P,R 134 | XP_016865173.1 |
Set Membership: |
HapMap |