Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607071 MIM: 604038 MIM: 602725 MIM: 607073 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HYAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HYAL1 - hyaluronoglucosaminidase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HYAL3 - hyaluronoglucosaminidase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001200029.1 | Intron | NP_001186958.1 | ||||
NM_001200030.1 | Intron | NP_001186959.1 | ||||
NM_001200031.1 | Intron | NP_001186960.1 | ||||
NM_001200032.1 | Intron | NP_001186961.1 | ||||
NM_003549.3 | Intron | NP_003540.2 |
IFRD2 - interferon related developmental regulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006764.4 | Intron | NP_006755.4 |
LSMEM2 - leucine rich single-pass membrane protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAT6 - N-acetyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |