Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601266 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUT PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DUT - deoxyuridine triphosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025248.1 | 45 | Intron | NP_001020419.1 | |||
NM_001025249.1 | 45 | Intron | NP_001020420.1 | |||
NM_001948.3 | 45 | UTR 5 | NP_001939.1 | |||
XM_011521315.2 | 45 | Intron | XP_011519617.1 | |||
XM_017021988.1 | 45 | Intron | XP_016877477.1 |
Set Membership: |
HapMap |