Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616797 MIM: 176830 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EFR3B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
EFR3B - EFR3 homolog B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POMC - proopiomelanocortin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000939.3 | 989 | Silent Mutation | NP_000930.1 | |||
NM_001035256.2 | 989 | Silent Mutation | NP_001030333.1 | |||
NM_001319204.1 | 989 | Silent Mutation | NP_001306133.1 | |||
NM_001319205.1 | 989 | Silent Mutation | NP_001306134.1 |