Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600956 MIM: 189906 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMHR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMHR2 - anti-Mullerian hormone receptor type 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164690.1 | 82 | Intron | NP_001158162.1 | |||
NM_001164691.1 | 82 | Intron | NP_001158163.1 | |||
NM_020547.2 | 82 | Intron | NP_065434.1 | |||
XM_011538173.1 | 82 | Intron | XP_011536475.1 | |||
XM_011538174.1 | 82 | Intron | XP_011536476.1 | |||
XM_011538176.1 | 82 | Intron | XP_011536478.1 | |||
XM_011538178.1 | 82 | Intron | XP_011536480.1 | |||
XM_011538179.1 | 82 | Intron | XP_011536481.1 | |||
XM_011538180.1 | 82 | Intron | XP_011536482.1 | |||
XM_011538181.1 | 82 | Intron | XP_011536483.1 | |||
XM_011538183.2 | 82 | Intron | XP_011536485.1 | |||
XM_011538184.2 | 82 | Intron | XP_011536486.1 | |||
XM_011538185.1 | 82 | Intron | XP_011536487.1 | |||
XM_011538186.2 | 82 | UTR 5 | XP_011536488.1 | |||
XM_011538187.2 | 82 | Intron | XP_011536489.1 | |||
XM_017019179.1 | 82 | Intron | XP_016874668.1 |
SP1 - Sp1 transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |