Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAACCCCTCCCGGGTTTTGCGGG[A/G]CTCCCGGGCGGCGCTCGGCTTTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123864 MIM: 604886 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX4I1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX4I1 - cytochrome c oxidase subunit 4I1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318786.1 | Intron | NP_001305715.1 | ||||
NM_001318788.1 | Intron | NP_001305717.1 | ||||
NM_001318794.1 | Intron | NP_001305723.1 | ||||
NM_001318797.1 | Intron | NP_001305726.1 | ||||
NM_001318802.1 | Intron | NP_001305731.1 | ||||
NM_001861.4 | Intron | NP_001852.1 |
EMC8 - ER membrane protein complex subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142288.1 | Intron | NP_001135760.1 | ||||
NM_006067.4 | Intron | NP_006058.1 | ||||
XM_011522813.2 | Intron | XP_011521115.1 | ||||
XM_017022867.1 | Intron | XP_016878356.1 |
LOC101928557 - uncharacterized LOC101928557 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |