Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609841 MIM: 609666 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6762 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR6762 - microRNA 6762 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC8B1 - solute carrier family 8 member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006719607.2 | 2377 | UTR 3 | XP_006719670.1 | |||
XM_011538749.2 | 2377 | UTR 3 | XP_011537051.1 | |||
XM_011538750.2 | 2377 | UTR 3 | XP_011537052.1 | |||
XM_011538752.2 | 2377 | UTR 3 | XP_011537054.1 | |||
XM_017019978.1 | 2377 | Intron | XP_016875467.1 |
TPCN1 - two pore segment channel 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |