Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147892 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DIO1 - deiodinase, iodothyronine type I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000792.6 | Intron | NP_000783.2 | ||||
NM_001039715.2 | Intron | NP_001034804.1 | ||||
NM_001039716.2 | Intron | NP_001034805.1 | ||||
NM_001324316.1 | Intron | NP_001311245.1 | ||||
NM_213593.4 | Intron | NP_998758.1 |
YIPF1 - Yip1 domain family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |