Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603372 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP128 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEP128 - centrosomal protein 128 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152446.3 | Intron | NP_689659.2 | ||||
XM_006720056.3 | Intron | XP_006720119.1 | ||||
XM_011536490.2 | Intron | XP_011534792.1 | ||||
XM_011536491.2 | Intron | XP_011534793.1 | ||||
XM_011536492.2 | Intron | XP_011534794.1 | ||||
XM_011536493.2 | Intron | XP_011534795.1 | ||||
XM_011536495.2 | Intron | XP_011534797.1 | ||||
XM_017021042.1 | Intron | XP_016876531.1 | ||||
XM_017021043.1 | Intron | XP_016876532.1 |
TSHR - thyroid stimulating hormone receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000369.2 | Intron | NP_000360.2 | ||||
NM_001018036.2 | Intron | NP_001018046.1 | ||||
NM_001142626.2 | Intron | NP_001136098.1 | ||||
XM_005268037.4 | Intron | XP_005268094.1 | ||||
XM_005268039.1 | Intron | XP_005268096.1 | ||||
XM_006720245.1 | Intron | XP_006720308.1 | ||||
XM_011537119.2 | Intron | XP_011535421.1 |
Set Membership: |
HapMap |