Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCATATTTTTAGTTGTGTGCCTA[A/G]CATAATTAATATGGTTGCTTTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610605 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPEB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPEB2 - cytoplasmic polyadenylation element binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177381.1 | Intron | NP_001170852.1 | ||||
NM_001177382.1 | Intron | NP_001170853.1 | ||||
NM_001177383.1 | Intron | NP_001170854.1 | ||||
NM_001177384.1 | Intron | NP_001170855.1 | ||||
NM_182485.2 | Intron | NP_872291.2 | ||||
NM_182646.2 | Intron | NP_872587.2 | ||||
XM_005248135.3 | Intron | XP_005248192.2 | ||||
XM_011513777.2 | Intron | XP_011512079.1 | ||||
XM_011513778.2 | Intron | XP_011512080.1 | ||||
XM_011513779.1 | Intron | XP_011512081.1 | ||||
XM_017007733.1 | Intron | XP_016863222.1 | ||||
XM_017007734.1 | Intron | XP_016863223.1 |
CPEB2-AS1 - CPEB2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101929095 - uncharacterized LOC101929095 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |