Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608140 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP35 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NUP35 - nucleoporin 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287584.1 | Intron | NP_001274513.1 | ||||
NM_001287585.1 | Intron | NP_001274514.1 | ||||
NM_138285.4 | Intron | NP_612142.2 | ||||
XM_006712254.2 | Intron | XP_006712317.1 | ||||
XM_011510576.2 | Intron | XP_011508878.1 | ||||
XM_011510577.2 | Intron | XP_011508879.1 | ||||
XM_017003308.1 | Intron | XP_016858797.1 |