Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RNU6-6P PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNU6-6P - RNA, U6 small nuclear 6, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UCMA - upper zone of growth plate and cartilage matrix associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303118.1 | 366 | Missense Mutation | TAT,TGT | Y,C 99 | NP_001290047.1 | |
NM_001303119.1 | 366 | Missense Mutation | TAT,TGT | Y,C 77 | NP_001290048.1 | |
NM_145314.2 | 366 | Missense Mutation | TAT,TGT | Y,C 131 | NP_660357.2 |