Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603889 MIM: 605466 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIRPB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SIRPB1 - signal regulatory protein beta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIRPG - signal regulatory protein gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039508.1 | 980 | UTR 3 | NP_001034597.1 | |||
NM_018556.3 | 980 | UTR 3 | NP_061026.2 | |||
NM_080816.2 | 980 | UTR 3 | NP_543006.2 | |||
XM_005260749.3 | 980 | Intron | XP_005260806.1 | |||
XM_011529286.2 | 980 | UTR 3 | XP_011527588.1 | |||
XM_011529287.2 | 980 | Intron | XP_011527589.1 |
SIRPG-AS1 - SIRPG antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |