Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194355 MIM: 612062 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
XBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
XBP1 - X-box binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079539.1 | Intron | NP_001073007.1 | ||||
NM_005080.3 | Intron | NP_005071.2 |
ZNRF3 - zinc and ring finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206998.1 | Intron | NP_001193927.1 | ||||
NM_032173.3 | Intron | NP_115549.2 | ||||
XM_011530435.2 | Intron | XP_011528737.1 | ||||
XM_011530436.2 | Intron | XP_011528738.1 | ||||
XM_011530437.1 | Intron | XP_011528739.1 | ||||
XM_011530438.2 | Intron | XP_011528740.1 | ||||
XM_017028990.1 | Intron | XP_016884479.1 |