Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611343 | ||||||||||||||||||||||||||||||||
Literature Links: |
IDNK PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
IDNK - IDNK, gluconokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001551.3 | 246 | Intron | NP_001001551.2 | |||
NM_001256915.1 | 246 | UTR 5 | NP_001243844.1 | |||
XM_006717110.3 | 246 | Intron | XP_006717173.1 | |||
XM_006717111.3 | 246 | Intron | XP_006717174.1 | |||
XM_017014719.1 | 246 | Intron | XP_016870208.1 | |||
XM_017014720.1 | 246 | Intron | XP_016870209.1 | |||
XM_017014721.1 | 246 | Intron | XP_016870210.1 | |||
XM_017014722.1 | 246 | Intron | XP_016870211.1 | |||
XM_017014723.1 | 246 | Intron | XP_016870212.1 | |||
XM_017014724.1 | 246 | UTR 5 | XP_016870213.1 | |||
XM_017014725.1 | 246 | Intron | XP_016870214.1 |
Set Membership: |
HapMap |