Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
66 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609831 MIM: 176763 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC163P PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC163P - coiled-coil domain containing 163, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMACHC - methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015506.2 | 379 | Silent Mutation | GTA,GTG | V,V 107 | NP_056321.2 | |
XM_005270724.4 | 379 | Silent Mutation | GTA,GTG | V,V 42 | XP_005270781.1 | |
XM_011541204.2 | 379 | Silent Mutation | GTA,GTG | V,V 50 | XP_011539506.1 |
PRDX1 - peroxiredoxin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |