Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGATGGGGCGTGGCTGTGTCAC[C/G]AAGGGCGTGGTCATGGAGTAGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605418 MIM: 601729 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DKKL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DKKL1 - dickkopf like acrosomal protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197301.1 | Intron | NP_001184230.1 | ||||
NM_001197302.1 | Intron | NP_001184231.1 | ||||
NM_014419.3 | Intron | NP_055234.1 | ||||
XM_006723142.2 | Intron | XP_006723205.1 | ||||
XM_011526724.1 | Intron | XP_011525026.1 | ||||
XM_011526725.1 | Intron | XP_011525027.1 | ||||
XM_011526726.2 | Intron | XP_011525028.1 | ||||
XM_011526727.2 | Intron | XP_011525029.1 | ||||
XM_011526728.2 | Intron | XP_011525030.1 | ||||
XM_011526729.1 | Intron | XP_011525031.1 | ||||
XM_017026593.1 | Intron | XP_016882082.1 |
LOC101928295 - uncharacterized LOC101928295 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEAD2 - TEA domain transcription factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |