Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613880 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BAHD1 - bromo adjacent homology domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301132.1 | Intron | NP_001288061.1 | ||||
NM_014952.4 | Intron | NP_055767.3 | ||||
XM_011521366.2 | Intron | XP_011519668.2 | ||||
XM_011521367.2 | Intron | XP_011519669.2 | ||||
XM_011521368.2 | Intron | XP_011519670.2 | ||||
XM_011521369.2 | Intron | XP_011519671.1 |