Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C18orf8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C18orf8 - chromosome 18 open reading frame 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276342.1 | Intron | NP_001263271.1 | ||||
NM_001318707.1 | Intron | NP_001305636.1 | ||||
NM_001318708.1 | Intron | NP_001305637.1 | ||||
NM_001318709.1 | Intron | NP_001305638.1 | ||||
NM_013326.4 | Intron | NP_037458.3 | ||||
XM_011525939.1 | Intron | XP_011524241.1 | ||||
XM_017025717.1 | Intron | XP_016881206.1 |
NPC1 - NPC intracellular cholesterol transporter 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000271.4 | Intron | NP_000262.2 | ||||
XM_005258277.1 | Intron | XP_005258334.1 | ||||
XM_005258278.4 | Intron | XP_005258335.1 | ||||
XM_005258279.2 | Intron | XP_005258336.1 | ||||
XM_006722479.3 | Intron | XP_006722542.1 | ||||
XM_011526015.1 | Intron | XP_011524317.1 | ||||
XM_017025784.1 | Intron | XP_016881273.1 | ||||
XM_017025785.1 | Intron | XP_016881274.1 | ||||
XM_017025786.1 | Intron | XP_016881275.1 | ||||
XM_017025787.1 | Intron | XP_016881276.1 |