Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602341 MIM: 614085 MIM: 604730 | ||||||||||||||||||||||||||||||||
Literature Links: |
FOXM1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
FOXM1 - forkhead box M1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHNO1 - RAD9-HUS1-RAD1 interacting nuclear orphan 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252499.2 | Intron | NP_001239428.1 | ||||
NM_001252500.2 | Intron | NP_001239429.1 | ||||
NM_001257097.1 | Intron | NP_001244026.1 | ||||
NM_001257098.1 | Intron | NP_001244027.1 |
TULP3 - tubby like protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |