Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATAAAACAAGAGGATATACATCC[A/G]CTTACATAAAACGAGAAATATGTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609803 MIM: 610073 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKAR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKAR - ankyrin and armadillo repeat containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORMDL1 - ORMDL sphingolipid biosynthesis regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128150.1 | 1161 | UTR 3 | NP_001121622.1 | |||
NM_016467.4 | 1161 | UTR 3 | NP_057551.1 | |||
XM_005246970.2 | 1161 | UTR 3 | XP_005247027.1 | |||
XM_011512199.2 | 1161 | Intron | XP_011510501.1 | |||
XM_011512200.2 | 1161 | Intron | XP_011510502.1 | |||
XM_017005347.1 | 1161 | UTR 3 | XP_016860836.1 | |||
XM_017005348.1 | 1161 | Intron | XP_016860837.1 |
OSGEPL1 - O-sialoglycoprotein endopeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSGEPL1-AS1 - OSGEPL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |