Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602103 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRAPPC10 - trafficking protein particle complex 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003274.4 | Intron | NP_003265.3 | ||||
XM_005261168.4 | Intron | XP_005261225.1 | ||||
XM_011529714.2 | Intron | XP_011528016.1 | ||||
XM_011529715.2 | Intron | XP_011528017.1 | ||||
XM_011529716.2 | Intron | XP_011528018.1 | ||||
XM_011529717.2 | Intron | XP_011528019.1 | ||||
XM_011529718.2 | Intron | XP_011528020.1 | ||||
XM_011529719.2 | Intron | XP_011528021.1 | ||||
XM_011529720.2 | Intron | XP_011528022.1 | ||||
XM_011529721.2 | Intron | XP_011528023.1 | ||||
XM_011529722.2 | Intron | XP_011528024.1 | ||||
XM_011529723.2 | Intron | XP_011528025.1 | ||||
XM_011529725.2 | Intron | XP_011528027.1 | ||||
XM_011529726.2 | Intron | XP_011528028.1 | ||||
XM_011529727.2 | Intron | XP_011528029.1 | ||||
XM_011529728.2 | Intron | XP_011528030.1 | ||||
XM_011529729.2 | Intron | XP_011528031.1 | ||||
XM_011529730.2 | Intron | XP_011528032.1 | ||||
XM_017028454.1 | Intron | XP_016883943.1 | ||||
XM_017028455.1 | Intron | XP_016883944.1 |