Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300014 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP2B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP2B3 - ATPase plasma membrane Ca2+ transporting 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001344.2 | Intron | NP_001001344.1 | ||||
NM_021949.3 | Intron | NP_068768.2 | ||||
XM_005274689.3 | Intron | XP_005274746.1 | ||||
XM_005274690.3 | Intron | XP_005274747.1 | ||||
XM_005274691.3 | Intron | XP_005274748.1 | ||||
XM_005274692.3 | Intron | XP_005274749.1 | ||||
XM_011531174.2 | Intron | XP_011529476.1 | ||||
XM_011531177.2 | Intron | XP_011529479.1 | ||||
XM_017029553.1 | Intron | XP_016885042.1 |