Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608959 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPH3 - diphthamide biosynthesis 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001047434.2 | 112 | Intron | NP_001040899.1 | |||
NM_206831.2 | 112 | Intron | NP_996662.1 |
OXNAD1 - oxidoreductase NAD binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138381.3 | 112 | UTR 5 | NP_612390.1 | |||
XM_005265559.3 | 112 | Intron | XP_005265616.1 | |||
XM_006713405.1 | 112 | UTR 5 | XP_006713468.1 | |||
XM_006713406.2 | 112 | UTR 5 | XP_006713469.1 | |||
XM_011534230.2 | 112 | Intron | XP_011532532.1 | |||
XM_011534232.2 | 112 | Intron | XP_011532534.1 | |||
XM_011534233.2 | 112 | Intron | XP_011532535.1 | |||
XM_011534234.2 | 112 | Intron | XP_011532536.1 | |||
XM_011534235.2 | 112 | Intron | XP_011532537.1 | |||
XM_017007485.1 | 112 | UTR 5 | XP_016862974.1 | |||
XM_017007486.1 | 112 | UTR 5 | XP_016862975.1 | |||
XM_017007487.1 | 112 | UTR 5 | XP_016862976.1 | |||
XM_017007488.1 | 112 | UTR 5 | XP_016862977.1 | |||
XM_017007489.1 | 112 | UTR 5 | XP_016862978.1 | |||
XM_017007490.1 | 112 | UTR 5 | XP_016862979.1 | |||
XM_017007491.1 | 112 | UTR 5 | XP_016862980.1 | |||
XM_017007492.1 | 112 | Intron | XP_016862981.1 | |||
XM_017007493.1 | 112 | UTR 5 | XP_016862982.1 | |||
XM_017007494.1 | 112 | UTR 5 | XP_016862983.1 | |||
XM_017007495.1 | 112 | UTR 5 | XP_016862984.1 | |||
XM_017007496.1 | 112 | UTR 5 | XP_016862985.1 |