Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608959 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPH3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DPH3 - diphthamide biosynthesis 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001047434.2 | 100 | Intron | NP_001040899.1 | |||
NM_206831.2 | 100 | Intron | NP_996662.1 |
OXNAD1 - oxidoreductase NAD binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138381.3 | 100 | UTR 5 | NP_612390.1 | |||
XM_005265559.3 | 100 | Intron | XP_005265616.1 | |||
XM_006713405.1 | 100 | UTR 5 | XP_006713468.1 | |||
XM_006713406.2 | 100 | UTR 5 | XP_006713469.1 | |||
XM_011534230.2 | 100 | Intron | XP_011532532.1 | |||
XM_011534232.2 | 100 | Intron | XP_011532534.1 | |||
XM_011534233.2 | 100 | Intron | XP_011532535.1 | |||
XM_011534234.2 | 100 | Intron | XP_011532536.1 | |||
XM_011534235.2 | 100 | Intron | XP_011532537.1 | |||
XM_017007485.1 | 100 | UTR 5 | XP_016862974.1 | |||
XM_017007486.1 | 100 | UTR 5 | XP_016862975.1 | |||
XM_017007487.1 | 100 | UTR 5 | XP_016862976.1 | |||
XM_017007488.1 | 100 | UTR 5 | XP_016862977.1 | |||
XM_017007489.1 | 100 | UTR 5 | XP_016862978.1 | |||
XM_017007490.1 | 100 | UTR 5 | XP_016862979.1 | |||
XM_017007491.1 | 100 | UTR 5 | XP_016862980.1 | |||
XM_017007492.1 | 100 | Intron | XP_016862981.1 | |||
XM_017007493.1 | 100 | UTR 5 | XP_016862982.1 | |||
XM_017007494.1 | 100 | UTR 5 | XP_016862983.1 | |||
XM_017007495.1 | 100 | UTR 5 | XP_016862984.1 | |||
XM_017007496.1 | 100 | UTR 5 | XP_016862985.1 |
Set Membership: |
HapMap |