Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612490 MIM: 616757 MIM: 611594 MIM: 607029 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf68 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf68 - chromosome 2 open reading frame 68 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF181 - ring finger protein 181 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016494.3 | 404 | Silent Mutation | TAC,TAT | Y,Y 118 | NP_057578.1 | |
XM_005264359.3 | 404 | Missense Mutation | ACG,ATG | T,M 132 | XP_005264416.1 |
TMEM150A - transmembrane protein 150A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP39 - ubiquitin specific peptidase 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VAMP5 - vesicle associated membrane protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |