Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609212 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASRGL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASRGL1 - asparaginase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083926.1 | 198 | UTR 5 | NP_001077395.1 | |||
NM_025080.3 | 198 | UTR 5 | NP_079356.3 | |||
XM_005274305.3 | 198 | UTR 5 | XP_005274362.1 | |||
XM_005274306.2 | 198 | UTR 5 | XP_005274363.1 | |||
XM_011545265.2 | 198 | UTR 5 | XP_011543567.1 | |||
XM_011545266.2 | 198 | UTR 5 | XP_011543568.1 | |||
XM_017018354.1 | 198 | UTR 5 | XP_016873843.1 |